missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, ACS Reagent, Anhydrous Granular, Honeywell Fluka™

ACS Reagent, anhydrous, granular,≥99.0%

41.84 € - 618.50 €

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15652450
View Documents
Honeywell-Fluka
239313-500G
500 g
41.84 €
500g
Please to purchase this item. Need a web account? Register with us today!
15622450
View Documents
Honeywell-Fluka
239313-1KG
1 kg
61.04 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15632450
View Documents
Honeywell-Fluka
239313-2.5KG
2.5 kg
134.45 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15662450
View Documents
Honeywell-Fluka
239313-5KG
5 kg
428.95 €
5kg
Please to purchase this item. Need a web account? Register with us today!
15642450
View Documents
Honeywell-Fluka
239313-4X2.5KG
4 x 2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15612450
View Documents
Honeywell-Fluka
239313-12KG
12 kg
533.46 €
12kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149

Specifications

7757-82-6
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99%
2.68g/mL at 25°C
Granules
Na2SO4
NONH for all modes of transport
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04g/mol
ACS Reagent
Sodium Sulfate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-820-9

missing translation for 'rtecsNumber' : WE1650000

SDS
Documents

Documents

Product Certifications