missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
12 kg
Unit Size:
12kg
1kg
2.5kg
25g
500g
5kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Anhydrous Powder |
| Quantity | 1 kg |
| Linear Formula | Na2SO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 142.04g/mol |
| Percent Purity | ≥99% |
| Grade | ACS Reagent |
| Density | 2.68g/mL at 25°C |
| Chemical Name or Material | Sodium Sulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction