missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical

Brand:  Fisher Chemical S/6650/60

21.79 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10746292

  • 31.50 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

1kg, Plastic powder jar

TRUSTED_SUSTAINABILITY
Specifications

Specifications

884°C
White
Solid
Na2O4S
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
Plastic powder jar
7757-82-6
5 to 8
1 kg
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical > 1kg, Plastic powder jar

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.