Learn More
Sodium sulfate decahydrate, 99%, Thermo Scientific Chemicals
52.56 € - 522.60 €
Chemical Identifiers
CAS | 7727-73-3 |
---|---|
Molecular Formula | H20Na2O14S |
Molecular Weight (g/mol) | 322.186 |
MDL Number | MFCD00149189 |
InChI Key | RSIJVJUOQBWMIM-UHFFFAOYSA-L |
Synonym | sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o |
PubChem CID | 62649 |
ChEBI | CHEBI:32586 |
IUPAC Name | disodium;sulfate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457667
|
Thermo Scientific Alfa Aesar
A15702.0B |
1000 g |
52.56 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467667
|
Thermo Scientific Alfa Aesar
A15702.0I |
5000 g |
165.83 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477667
|
Thermo Scientific Alfa Aesar
A15702.0G |
25,000 g |
522.60 €
25000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.
Solubility
Soluble in water and glycerin. Insoluble in ethanol and methanol.
Notes
Stable. Incompatible with strong acids, aluminum and magnesium.
Chemical Identifiers
7727-73-3 | |
322.186 | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
62649 | |
disodium;sulfate;decahydrate |
H20Na2O14S | |
MFCD00149189 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
CHEBI:32586 | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
∼32°C | |
5.2 to 8.0 | |
MFCD00149189 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
disodium;sulfate;decahydrate | |
62649 | |
322.19 (142.04 Anhydrous) | |
Odorless | |
Sodium sulfate decahydrate |
7727-73-3 | |
H20Na2O14S | |
14,8680 | |
Soluble in water and glycerin. Insoluble in ethanol and methanol. | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
322.186 | |
CHEBI:32586 | |
99% | |
1.46 |
Safety and Handling
missing translation for 'einecsNumber' : 231-820-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.