missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Honeywell

Purum p.a., anhydrous,≥99.0% (T)

40.59 € - 173.33 €

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15666870
View Documents Promotion Details
Honeywell Research Chemicals
71962-1KG
1 kg
40.59 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15676870
View Documents Promotion Details
Honeywell Research Chemicals
71962-5KG
5 kg
153.65 €
5kg
Please to purchase this item. Need a web account? Register with us today!
15686870
View Documents Promotion Details
Honeywell Research Chemicals
71962-6X1KG
6 x 1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149

Specifications

7757-82-6
Na2SO4
NONH for all modes of transport
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04g/mol
2.68g/mL at 25°C
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99% (T)
Sodium sulfate
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : WE1650000

SDS
Documents

Documents

Product Certifications