missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals

Product Code. 10618264
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-82-2
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00003498
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym sodum sulfuret, sodium sulfide, disodium sulphide, sodium sulfide van, na2-s, disodium monosulfide, na2s, sodium sulfide, anhydrous dot, hsdb 772, sodium sulfide na2s
PubChem CID 237873
SMILES [Na+].[Na+].[S--]

Specifications

Color Beige-Yellow
Physical Form Lump/Powder
Quantity 50 g
Assay Percent Range 90% min. (Iodimetry)
Linear Formula Na2S
Merck Index 15,881
Solubility Information Solubility aq. soln.: 186 g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether
IUPAC Name disodium sulfanide
Formula Weight 78.04
Percent Purity ≥90%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium sulfide, 90%, balance water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'