missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10656811

Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10656811
25 g, Glass bottle
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10656811

Brand: Thermo Scientific Acros 424420250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Melting Point 50.0°C
Color Colorless or Yellow
Physical Form Adhering Crystals Chunks
Quantity 25 g
Assay Percent Range 98% min. (Iodimetry)
Linear Formula Na2S·9H2O
Fieser 01,1104; 11,492
Merck Index 15, 8810
Solubility Information Solubility in water: 180g/L (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 240.18
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Sodium sulfide nonahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462729 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number424425000,424420250,424420025
Lot NumberA0439254 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number424425000,424420250,424420025
Lot NumberA0459803 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number424425000,424420250,424420025
Lot NumberA0437124 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number424425000,424420250,424420025
Lot NumberB0154446 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number424420250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium sulfide nonahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H290-May be corrosive to metals.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.