missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Nonahydrate, Honeywell Fluka™

ACS Reagent,≥98.0%

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15611820
View Documents Promotion Details
Honeywell-Fluka
208043-5G
5 g N/A
N/A
15681810
View Documents Promotion Details
Honeywell-Fluka
208043-25G
25 g N/A
N/A
15671810
View Documents Promotion Details
Honeywell-Fluka
208043-100G
100 g N/A
N/A
15601820
View Documents Promotion Details
Honeywell-Fluka
208043-500G
500 g N/A
N/A
15691810
View Documents Promotion Details
Honeywell-Fluka
208043-3KG
3 kg N/A
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1313-84-4
240.17
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
H18Na2O9S
MFCD00149184
disodium nonahydrate sulfanediide

Specifications

1313-84-4
H18Na2O9S
MFCD00149184
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
disodium nonahydrate sulfanediide
240.18g/mol
ACS Reagent
Colorless or Yellow
Na2S·9H2O
1849
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
240.17
≥98%
Sodium Sulfide Nonahydrate
Videos
Safety and Handling

Safety and Handling

P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391

H302-H311-H314-H400

missing translation for 'einecsNumber' : 215-211-5

missing translation for 'rtecsNumber' : WE1925000

Product Identifier
  • Sodium sulfide nonahydrate

Signal Word
  • Danger

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : disodium sulphide

SDS
Documents

Documents

Product Certifications