missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 97%, extra pure, anhydrous, Thermo Scientific Chemicals

46.43 € - 266.33 €

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10286390
View Documents
Thermo Scientific Acros
196630010
1 kg Plastic Bottle
46.43 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10308050
View Documents
Thermo Scientific Acros
196630025
2.5 kg Plastic Bottle
91.66 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11904231
View Documents
Thermo Scientific Acros
196630100
10 kg Plastic drum
266.33 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

7757-83-7
126.04
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
CHEBI:86477
[O-]S(=O)[O-].[Na+].[Na+]
Na2O3S
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
disodium;sulfite

Specifications

>500.0°C
White to Yellow
Fine Crystals or Crystalline Powder
Na2O3S
02,387
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
GEHJYWRUCIMESM-UHFFFAOYSA-L
disodium;sulfite
24437
126.04
Extra Pure
2.6300g/mL
7757-83-7
8.5 to 10.5 (5% soln.)
96% min. (Iodimetry)
Na2SO3
15,8811
Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
[O-]S(=O)[O-].[Na+].[Na+]
126.04
CHEBI:86477
97%
2.63
Sodium sulfite
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-821-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.