missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite Anhydrous, Certified AR for Analysis, Fisher Chemical™
34.35 € - 62.50 €
Chemical Identifiers
CAS | 7757-83-7 |
---|---|
Molecular Formula | Na2O3S |
Molecular Weight (g/mol) | 126.037 |
MDL Number | 3503 |
InChI Key | GEHJYWRUCIMESM-UHFFFAOYSA-L |
Synonym | sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
PubChem CID | 24437 |
ChEBI | CHEBI:86477 |
IUPAC Name | disodium;sulfite |
SMILES | [O-]S(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10070400
|
Fisher Chemical
S/6850/53 |
500 g |
34.35 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10704391
|
Fisher Chemical
S/6850/60 |
1 kg |
62.50 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7757-83-7 | |
126.037 | |
GEHJYWRUCIMESM-UHFFFAOYSA-L | |
24437 | |
disodium;sulfite |
Na2O3S | |
3503 | |
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
CHEBI:86477 | |
[O-]S(=O)[O-].[Na+].[Na+] |
Specifications
500°C | |
White | |
Solid | |
3503 | |
GEHJYWRUCIMESM-UHFFFAOYSA-L | |
disodium;sulfite | |
24437 | |
126.04g/mol |
7757-83-7 | |
8.5 to 10 | |
Na2O3S | |
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
[O-]S(=O)[O-].[Na+].[Na+] | |
126.037 | |
CHEBI:86477 | |
Plastic powder jar |