missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Solution, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11494739
Click to view available options
Quantity:
100 mL
Packaging:
Tamper evident
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Sodium Sulfide Solution, EP Grade, Reagecon™ is a ready-to-use reagent with 12g sodium sulfide diluted to 100mL with an aqueous glycerol solution for use in manual and automated titrations.

This sodium sulfide solution is prepared by dissolving 12g of sodium sulfide in 45mL solvent mixture of 10 parts water and 29 parts glycerol (85%), and diluted with that same mixture to 100mL.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

CAS 1313-84-4, 56-81-5, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 100 mL
Molecular Formula H18Na2O9S
MDL Number MFCD00149184
Synonym Disodium sulphide nonahydrate
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
IUPAC Name disodium nonahydrate sulfanediide
Molecular Weight (g/mol) 240.17
Formula Weight 240.2g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Concentration or Composition (by Analyte or Components) 12g/100mL
Odor Hydrogen sulfide (rotten eggs)
Packaging Tamper evident
Chemical Name or Material Sodium Sulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • SODIUM SULPHIDE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sodium sulphide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'