Learn More
Sodium tellurate(VI) hydrate, Te 45% min, Thermo Scientific Chemicals
Description
Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | (decomposition) |
Physical Form | Crystalline |
Quantity | 10 g |
Merck Index | 14,8686 |
Solubility Information | Soluble in water. |
IUPAC Name | disodium tellurate |
Formula Weight | 237.58 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | Te 45% min |
Chemical Name or Material | Sodium tellurate (VI) hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.