Learn More
Sodium tert-butoxide, 99%, pure, Thermo Scientific Chemicals
30.80 € - 694.00 €
Chemical Identifiers
CAS | 865-48-5 |
---|---|
Molecular Formula | C4H9NaO |
Molecular Weight (g/mol) | 96.1 |
MDL Number | MFCD00040575 |
InChI Key | MFRIHAYPQRLWNB-UHFFFAOYSA-N |
Synonym | sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide |
PubChem CID | 23676156 |
IUPAC Name | sodium;2-methylpropan-2-olate |
SMILES | CC(C)(C)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10728362
|
Thermo Scientific Acros
252550050 |
5 g | Glass bottle |
30.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247350
|
Thermo Scientific Acros
252551000 |
100 g | Glass bottle |
53.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10723924
|
Thermo Scientific Acros
252555000 |
500 g | Glass bottle |
174.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905821
|
Thermo Scientific Acros
252550025 |
2.5 kg | Plastic Drum |
694.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
865-48-5 | |
96.1 | |
MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
23676156 | |
CC(C)(C)[O-].[Na+] |
C4H9NaO | |
MFCD00040575 | |
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
sodium;2-methylpropan-2-olate |
Specifications
865-48-5 | |
12°C | |
C4H9NaO | |
MFCD00040575 | |
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
sodium;2-methylpropan-2-olate | |
23676156 | |
99% | |
Sodium tert-butoxide |
180.0°C | |
99% | |
NaOC(CH3)3 | |
16,55 | |
Solubility in water: reacts. | |
CC(C)(C)[O-].[Na+] | |
96.1 | |
96.1 | |
Pure |
Safety and Handling
GHS H Statement
Self-heating; may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
Flammable solid.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-741-9
- Sodium tert-butoxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H251-Self-heating: may catch fire.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only