missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals
31.05 € - 299.49 €
Chemical Identifiers
CAS | 1330-43-4 |
---|---|
Molecular Formula | B4Na2O7 |
Molecular Weight (g/mol) | 201.21 |
MDL Number | MFCD00081185 |
InChI Key | NXSVMNYRVBQCMO-UHFFFAOYSA-N |
Synonym | Borax, fused |
IUPAC Name | disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
SMILES | [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10378100
|
Thermo Scientific Acros
206291000 |
100 g | Plastic bottle |
31.05 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042780
|
Thermo Scientific Acros
206295000 |
500 g | Plastic bottle |
91.05 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904331
|
Thermo Scientific Acros
206290025 |
2.5 kg | Plastic bottle |
299.49 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1330-43-4 | |
201.21 | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
B4Na2O7 | |
MFCD00081185 | |
Borax, fused | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
Specifications
1575.0°C | |
1330-43-4 | |
Crystalline Powder and/or Crystals | |
B4Na2O7 | |
MFCD00081185 | |
15,8726 | |
Solubility in water: 26g/L (20°C). Other solubilities: insoluble in alcohol | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
201.21 | |
98% | |
Sodium tetraborate |
741.0°C | |
Orange | |
98% | |
Na2B4O7 | |
04,461; 15,295 | |
Borax, fused | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) | |
201.22 | |
Analytical |
Safety and Handling
GHS H Statement
May damage fertility.
May damage the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-540-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium tetraborate, anhydrous
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only