missing translation for 'onlineSavingsMsg'
Learn More
Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10577562

Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10577562
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10577562

Brand: Thermo Scientific Acros 419450250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Molecular Weight (g/mol) 381.36
MDL Number MFCD00149193
Synonym Borax, Sodium borate decahydrate

Specifications

Melting Point 75.0°C
Color White
pH 9.15 to 9.20 (0.01M soln. at 25°C)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.5+%
Linear Formula Na2B4O7·10H2O
Fieser 04,461
Merck Index 15, 8726
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 381.36
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468895 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number419450025,419450250,419450010
Lot NumberA0465470 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number419450025,419450250,419450010
Lot NumberA0465491 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number419450025,419450250,419450010
Lot NumberA0462507 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number419450025,419450250,419450010
Lot NumberA0458123 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number419450025,419450250,419450010
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.