missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
Especificaciones
Especificaciones
Melting Point | 75.0°C |
Color | White |
pH | 9 to 9.5 (5% soln. at 20°C) |
Physical Form | Fine Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 99.5% |
Linear Formula | Na2B4O7·10H2O |
Fieser | 04,461 |
Merck Index | 15,8726 |
Solubility Information | Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'