missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 6 x 500 g |
Linear Formula | Na2B4O7 · 10H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 381.37g/mol |
Percent Purity | ≥99.5% |
Density | 1.73g/mL at 25°C |
Chemical Name or Material | Sodium tetraborate decahydrate |
Safety and Handling
Product Identifier
- Sodium tetraborate decahydrate
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction