Learn More
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water, Thermo Scientific Chemicals
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, C32H12BF24Na, CAS Number-79060-88-1
141.00 € - 1424.00 €
Chemical Identifiers
CAS | 79060-88-1 |
---|---|
Molecular Formula | C32H12BF24Na |
Molecular Weight (g/mol) | 886.209 |
MDL Number | MFCD00043323 |
InChI Key | LTGMONZOZHXAHO-UHFFFAOYSA-N |
Synonym | sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1- |
PubChem CID | 23681909 |
IUPAC Name | sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide |
SMILES | [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15475125
|
Thermo Scientific Alfa Aesar
H30014.03 |
1 g |
141.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15485125
|
Thermo Scientific Alfa Aesar
H30014.06 |
5 g |
454.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495125
|
Thermo Scientific Alfa Aesar
H30014.14 |
25 g |
1424.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
79060-88-1 | |
886.209 | |
LTGMONZOZHXAHO-UHFFFAOYSA-N | |
23681909 | |
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+] |
C32H12BF24Na | |
MFCD00043323 | |
sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1- | |
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide |
Specifications
79060-88-1 | |
C32H12BF24Na | |
sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1- | |
LTGMONZOZHXAHO-UHFFFAOYSA-N | |
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide | |
23681909 | |
97% | |
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate |
310°C (decomposition) | |
MFCD00043323 | |
Soluble in water. | |
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+] | |
886.209 | |
886.2 | |
may contain 1 to 5% water |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Sodium tetrakis¢3,5-bis(trifluoromethyl)phenyl!borate, may contain 1-5% water
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only