missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiocyanate, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10266300
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium;thiocyanate
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287.0°C
Color White
pH 5.5 to 7.5 (5% soln. at 20°C)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 500 g
Assay Percent Range 97.5% min. (on dried substance) (Argentometry)
Linear Formula NaSCN
Fieser 01,1105
Merck Index 15, 9480
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Formula Weight 81.07
Percent Purity 98%
Grade Extra Pure
Loss on Drying 2.5% max.
Packaging Plastic bottle
Chemical Name or Material Sodium thiocyanate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'