missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Anhydrous, Extra Pure, SLR, Fisher Chemical

Product Code. 10744961
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 287°C
CAS 540-72-7
Color White
pH 5.5 to 7.5
Physical Form Solid
Quantity 500 g
Molecular Formula CNNaS
MDL Number MFCD00011123
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES [Na+].[S-]C#N
IUPAC Name sodium cyanosulfanide
Molecular Weight (g/mol) 81.07
PubChem CID 516871
ChEBI CHEBI:30952
Formula Weight 81.07g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium thiocyanate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.
  • P233-Keep container tightly closed.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.