missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 90+%, technical, Thermo Scientific Chemicals

Product Code. 17251336
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.08
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
IUPAC Name sodium methylsulfanide
SMILES [Na+].C[S-]

Specifications

Assay Percent Range 90+%
Linear Formula CH3SNa
Quantity 1 g
Solubility Information Solubility aq. soln.: freely soluble.
Formula Weight 70.09
Grade Technical
Chemical Name or Material Sodium thiomethoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.