missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals

38.89 € - 322.61 €

Specifications

Chemical Name or Material Sodium thiosulfate
CAS 7772-98-7, 7732-18-5
Color Colorless
Physical Form Liquid
Molecular Formula Na2O3S2
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10173600
View Documents
Thermo Scientific Acros
124270010
1 L Plastic bottle
38.89 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11944231
View Documents
Thermo Scientific Acros
124270025
2.5 L Plastic Bottle
50.85 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
13468338
View Documents
Thermo Scientific Acros
124270250
25 L Plastic Drum
322.61 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent, Volumetric standard solution
Specifications

Specifications

Sodium thiosulfate
Colorless
Na2O3S2
MFCD00003499
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
[Na+].[Na+].[O-]S([S-])(=O)=O
24477
0.0950 to 0.1050N (20°C)
7772-98-7, 7732-18-5
Liquid
Na2S2O3
15, 8821
AKHNMLFCWUSKQB-UHFFFAOYSA-L
158.10
158.11
Videos
Safety and Handling

Safety and Handling

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only