missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
20 L
Description
Sodium thiosulfate, 0.1N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Immiscible with alcohol. |
IUPAC Name | disodium sulfanidesulfonate |
Formula Weight | 158.1 |
Concentration or Composition (by Analyte or Components) | Sodium thiosulphate: 1.58%; Water: 98.42% |
Appearance | Clear liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Chemical Name or Material | Sodium thiosulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'