missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Product Code. 15213406
Click to view available options
Quantity:
250 g
1 kg
5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Melting Point 48.0°C
pH 6.0 to 8.4 (5% aq. soln.)
Quantity 250 g
Assay Percent Range 98.5% min. (Iodimetry)
Linear Formula Na2S2O3
Merck Index 15,8821
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.1
Percent Purity 99%
Grade Analytical
Loss on Drying 1% max. (105°C)
Chemical Name or Material Sodium thiosulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.