missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate concentrate, For 1L standard solution, 0.01 M Na2S2O3 (0.01N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 Pc. |
Linear Formula | Na2S2O3 |
UN Number | NONH for all modes of transport |
Formula Weight | 158.11g/mol |
Packaging | Ampoule |
Chemical Name or Material | Sodium thiosulfate concentrate |
Safety and Handling
Product Identifier
- Sodium thiosulfate concentrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'