missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, 99+%, extra pure, Thermo Scientific Chemicals
34.07 € - 334.67 €
Chemical Identifiers
CAS | 10102-17-7 |
---|---|
Molecular Formula | Na2O3S2·5H2O |
Molecular Weight (g/mol) | 248.18 |
MDL Number | MFCD00149186 |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10724071
|
Thermo Scientific Acros
123370010 |
1 kg | Plastic bottle |
34.07 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10604982
|
Thermo Scientific Acros
123370025 |
2.5 kg | Plastic bottle |
75.68 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10003573
|
Thermo Scientific Acros
123370250 |
25 kg | Fibre drum |
334.67 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
10102-17-7 | |
248.18 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Na2O3S2·5H2O | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Specifications
48.5°C | |
Colorless to White | |
Crystals | |
Na2O3S2·5H2O | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.18 | |
Extra Pure | |
1.7200g/mL |
10102-17-7 | |
6 to 8.4 (10% soln. at 20°C) | |
99.0 to 101% (Iodimetry) | |
Na2S2O3·5H2O | |
15, 8821 | |
Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in turpentine | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.18 | |
CHEBI:32150 | |
≥99% | |
1.72 | |
Sodium thiosulfate pentahydrate |
RUO – Research Use Only