missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 500g, CHEBI:32150, White
Specifications
Specifications
Melting Point | 48.5°C |
Color | White |
pH | 6.0 to 8.4 |
Physical Form | Solid |
Quantity | 3 kg |
Formula Weight | 248.18g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'