missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Max % | 100.0000% |
Quantity | 50 kg |
Linear Formula | CaCl2 · 2H2O |
MDL Number | MFCD00149613 |
UN Number | NONH for all modes of transport |
Formula Weight | 248.18 |
Percent Purity | >99.5% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Fiberboard Box |
Density | 1.73 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'