missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10243960
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Liquid
Quantity 2.5 L
Packaging HDPE plastic bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'