missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10070220
Click to view available options
Quantity:
1 AMP
6 AMP
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

MDL Number 3499

Specifications

Boiling Point 100°C
Melting Point 0°C
Color Colorless
Physical Form Liquid
Quantity 6 AMP
Packaging Ampoule
Vapor Pressure 14mmHg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.