missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate solution, Volumetric, Reag. Ph. Eur., 0.1 M Na2S2O3 (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1 M Na2S2O3 (0.1N)
40.86 € - 260.72 €
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
PubChem CID | 24477 |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15674870
|
Honeywell-Fluka
35245-1L |
1 L |
40.86 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694870
|
Honeywell-Fluka
35245-5L-VP |
5 L |
148.78 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684870
|
Honeywell-Fluka
35245-5L |
5 L | N/A | ||||||
15604880
|
Honeywell-Fluka
35245-6X1L |
6 x 1 L |
211.51 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664870
|
Honeywell-Fluka
35245-10L-VP |
10 L |
260.72 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654870
|
Honeywell-Fluka
35245-10L |
10 L | N/A | ||||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
24477 |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
7772-98-7 | |
Na2S2O3 | |
NONH for all modes of transport | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
24477 | |
Sodium thiosulfate solution |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.11g/mol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title