missing translation for 'onlineSavingsMsg'
Learn More
Sodium Thiosulphate Solution, 0.1M, Fisher Chemical™ Product Code.: 12983624

Sodium Thiosulphate Solution, 0.1M, Fisher Chemical™

Product Code. 12983624
1000 mL
Click to view available options
Quantity:
100 mL
1000 mL
This item is not returnable. View return policy

Product Code. 12983624

Brand: Fisher Chemical VL 311 1000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Concentration 0.1 mol/L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1104135 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberVL 311 1000
Lot Number1102100 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberVL 311 1000
Lot Number1099374 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberVL 311 1000
Lot Number1095482 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberVL 311 1000
Lot Number1088260 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberVL 311 1000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Thiosulphate Solution, 0.1M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.