missing translation for 'onlineSavingsMsg'
Learn More
Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals Product Code.: 10184970

Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals

Product Code. 10184970
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10184970

Brand: Thermo Scientific Acros 291820250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56553-60-7
Molecular Formula C6H10BNaO6
Molecular Weight (g/mol) 211.94
MDL Number MFCD00012211
InChI Key HHYFEYBWNZJVFQ-UHFFFAOYSA-N
Synonym c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
IUPAC Name sodium bis(acetyloxy)boranuidyl acetate
SMILES [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point 114°C to 118°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula NaB(O2CCH3)3H
Quantity 25 g
Fieser 12,453; 13,283; 06,553; 16,309
Merck Index 15, 8822
Solubility Information Solubility in water: reacts. Other solubilities: soluble in thf,acetonitrile,1,2-dichloroethane
Formula Weight 211.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Sodium triacetoxyborohydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465884 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number291825000,291820250,291821000
Lot NumberA0461245 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number291825000,291820250,291821000
Lot NumberA0454039 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number291825000,291820250,291821000
Lot NumberA0403459 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number291825000,291820250,291821000
Lot NumberA0400312 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number291825000,291820250,291821000
compliance-icons compliance-icons
Product Identifier
  • Sodium triacetoxyborohydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.