Learn More
Sodium trifluoroacetate, 98%, Thermo Scientific Chemicals
Description
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.

Specifications
Specifications
Melting Point | ∼207°C (decomposition) |
Assay Percent Range | 98% |
Quantity | 100 g |
Beilstein | 3597949 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
IUPAC Name | sodium trifluoroacetate |
Formula Weight | 136 |
Percent Purity | 98% |
Chemical Name or Material | Sodium trifluoroacetate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252612 | Certificate of Analysis | 03/10/2024 | A14613.14,A14613.36,A14613.22 |
10249695 | Certificate of Analysis | 16/02/2024 | A14613.14,A14613.36,A14613.22 |
10218941 | Certificate of Analysis | 22/11/2021 | A14613.14,A14613.36,A14613.22 |
10206756 | Certificate of Analysis | 22/11/2021 | A14613.14,A14613.36,A14613.22 |
10165270 | Certificate of Analysis | 22/11/2021 | A14613.14,A14613.36,A14613.22 |
Safety and Handling


- Sodium trifluoroacetate
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H300-Fatal if swallowed.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- MIXTURE LIST-Contains : Sodium trifluoroacetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.