missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

25.02 € - 694.46 €

Chemical Identifiers

CAS 10213-10-2
Molecular Formula Na2O42H2O
Molecular Weight (g/mol) 329.85
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10117423
View Documents
Thermo Scientific Acros
207620050
5 g Glass bottle
25.02 €
5g
Please to purchase this item. Need a web account? Register with us today!
10144740
View Documents
Thermo Scientific Acros
207621000
100 g Glass bottle
52.36 €
100g
Please to purchase this item. Need a web account? Register with us today!
10792821
View Documents
Thermo Scientific Acros
207625000
500 g Glass bottle
181.91 €
500g
Please to purchase this item. Need a web account? Register with us today!
13468458
View Documents
Thermo Scientific Acros
207620025
2.5 kg Plastic bottle
694.46 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
Specifications

Chemical Identifiers

10213-10-2
329.85
WPZFLQRLSGVIAA-UHFFFAOYSA-N
150191
disodium;dioxido(dioxo)tungsten;dihydrate
Na2O42H2O
MFCD00149190
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
CHEBI:63939
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

10213-10-2
Crystalline Flakes or Powder
Na2O42H2O
MFCD00149190
15,8826
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
329.85
CHEBI:63939
≥99%
4.18
White
99% min
Na2WO4·2H2O
15,295
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
WPZFLQRLSGVIAA-UHFFFAOYSA-N
disodium;dioxido(dioxo)tungsten;dihydrate
150191
329.85
Analytical
Sodium tungstate dihydrate, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Warning

missing translation for 'rtecsNumber' : YO7900000

Product Identifier
  • Sodium tungstate dihydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Sodium tungstate dihydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only