Learn More
Sodium tungsten oxide dihydrate, 95%, Thermo Scientific Chemicals
98.50 € - 287.00 €
Chemical Identifiers
CAS | 10213-10-2 |
---|---|
Molecular Formula | H4Na2O6W |
Molecular Weight (g/mol) | 329.846 |
MDL Number | MFCD00149190 |
InChI Key | WPZFLQRLSGVIAA-UHFFFAOYSA-N |
Synonym | sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o |
PubChem CID | 150191 |
ChEBI | CHEBI:63939 |
IUPAC Name | disodium;dioxido(dioxo)tungsten;dihydrate |
SMILES | O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334979
|
Thermo Scientific Alfa Aesar
087371.30 |
250 g |
98.50 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324979
|
Thermo Scientific Alfa Aesar
087371.A1 |
1 kg |
287.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In organic chemistry, sodium tungstate is used as catalyst for epoxidation of alkenes and oxidation of alcohols into aldehydes or ketones. Used in examining the serum protein, serum protein-free filtrate. Used as alkaloid precipitating agent. Used in manufacture of metal tungsten, tungstate and tungsten oxide. Waterproof and fireproof fabric.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10213-10-2 | |
329.846 | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
150191 | |
disodium;dioxido(dioxo)tungsten;dihydrate |
H4Na2O6W | |
MFCD00149190 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
CHEBI:63939 | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] |
Specifications
100°C (in -2H2O) | |
White | |
H4Na2O6W | |
14,8698 | |
Soluble in water; Insoluble in alcohol. | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] | |
329.846 | |
CHEBI:63939 | |
95% | |
3.250 g/mL |
10213-10-2 | |
Crystalline | |
MFCD00149190 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)tungsten;dihydrate | |
150191 | |
329.86 (293.83 Anhydrous) | |
Odorless | |
Sodium tungsten oxide dihydrate |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302
missing translation for 'einecsNumber' : 236-743-4
missing translation for 'rtecsNumber' : YO7900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only