missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sofosbuvir impurity D, MedChemExpress
MedChemExpress Sofosbuvir impurity D is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
2390.85 € - 5578.20 €
Specifications
Chemical Name or Material | Sofosbuvir impurity D |
---|---|
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C22H29FN3O9P |
SMILES | O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16471248
|
Medchem Express
HY-I0723 |
5 mg |
5578.20 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16432469
|
Medchem Express
HY-I0723 |
1 mg |
2390.85 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sofosbuvir impurity D is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.Specifications
Sofosbuvir impurity D | |
Research | |
O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 | |
529.45 | |
Solid |
Light Yellow | |
C22H29FN3O9P | |
529.45 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only