missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity I, MedChemExpress

MedChemExpress Sofosbuvir impurity I, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

2390.85 € - 5578.20 €

Specifications

Chemical Name or Material Sofosbuvir impurity I
CAS 2164516-85-0
Color White
Purity Grade Notes Research
Molecular Formula C21H27FN3O9P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16482459
Encompass_Preferred
View Documents
Medchem Express
HY-I0512
1 mg
2390.85 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16401248
Encompass_Preferred
View Documents
Medchem Express
HY-I0512
5 mg
5578.20 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sofosbuvir impurity I, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Specifications

Specifications

Sofosbuvir impurity I
White
C21H27FN3O9P
515.43
Research
COVID-19-anti-virus
2164516-85-0
Research
O=[P@@](N[C@@H](C)C(OCC)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
515.43
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only