Learn More
Sofosbuvir impurity K, MedChemExpress
MedChemExpress Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Brand:  Medchem Express HY-I0515
Encompass_Preferred
 View more versions of this product
Product Code. 16402469
  • 1757.70 € / 1mg
Explore more special offers
Description
Description
Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Specifications
Specifications
Sofosbuvir impurity K
Light Yellow
C22H29ClN3O9P
O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](Cl)(C)[C@H](N2C(NC(C=C2)=O)=O)O1)OC3=CC=CC=C3
545.91
Solid
1496552-51-2
Research
1 mg
545.91
Research
COVID-19-anti-virus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Sofosbuvir impurity K, MedChemExpress