Learn More
Memmert™ Software AtmoCONTROL for SN260plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SN260plus Sterilizer
Unit Size:
Each
Description
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
| Certifications/Compliance | Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) |
| For Use With (Equipment) | SN260plus Sterilizer |
| Product Type | Software |