Learn More
Thermo Scientific™ SOLAμ™ SPE Plates
Remove variability and achieve highly consistent results. High reproducibility of Thermo Scientific™ SOLAμ™ SPE plates deliver greater sample success at low elution volumes.
338.75 € - 339.06 €
Specifications
Bed Weight 2 mg
Product Line SOLAμ
Type 96-well SPE Plate
Volume (Metric) Well 1 mL
View More Specs
Products 5
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15261808
View Documents
Thermo Scientific™
60209-001
HRP 2 mg/1 mL 96 well plate
338.75 €
Each
In Stock
to see stock.
Add to basket
15271808
View Documents
Thermo Scientific™
60209-002
SCX 2 mg/1 mL 96 well plate
339.06 €
Each
In Stock
to see stock.
Add to basket
15281808
View Documents
Thermo Scientific™
60209-003
SAX 2 mg/1 mL 96 well plate
339.06 €
Each
In Stock
to see stock.
Add to basket
15291808
View Documents
Thermo Scientific™
60209-004
WCX 2 mg/1 mL 96 well plate
338.94 €
Each
In Stock
to see stock.
Add to basket
15201818
View Documents
Thermo Scientific™
60209-005
WAX 2 mg/1 mL 96 well plate
338.94 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Remove variability and optimize high throughput laboratory workflows. Thermo Scientific™ SOLAμ™ Solid Phase Extraction (SPE) Plates are designed for bioanalytical and clinical research analysts who require cleaner, highly reproducible, and robust sample extraction using very low sample and solvent volumes in high throughput workflows. SOLAμ Well Plates achieve this due to the unique and award winning innovative frit-less SPE technology.
Ideal for Bioanalytical and Clinical Research Laboratories
Bioanalytical laboratories continue to drive to higher efficacy drugs and long acting formulations which require lower quantification limits. There is also the desire to take advantage of the replacement, refinement and reduction policy which is resulting in reduced animal model use. The growth of biopharmaceuticals also brings into consideration additional analytical issues such as solvation and non-specific binding.
Meet Demanding Sample Preparation Requirements
  • Processing of low sample volumes
  • High sensitivity due to low elution volumes
  • High reproducibility
  • Low analytical sample failures
  • Ease of use
  • High throughput processing
SOLAμ is the first micro-elution product to meet the requirements of the bioanalyst in combination with a robust, easy to use format.
Specifications
Specifications
2 mg
SOLAμ
96-well SPE Plate
1 mL
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.