Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Brand:  Thermo Scientific™ SK1830D
 View more versions of this product
Product Code. 16201591
  • 1296.45 € / 1 unit
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Services
1 Year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dual Stack Universal Platform
18 x 30 in.
457.2 mm
610mm
Solaris 4000
1 Year
Certificate of Analysis (COA), Certificate of Conformance (COC)
18 in.
30 in.
Solaris 18x30 Dual Stack Universal platform
Two platforms, screws, and supporting rods