Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Brand:  Thermo Scientific™ SK1214DK
 View more versions of this product
Product Code. 16241581
  • 541.70 € / 1 unit
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Services
1 Year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dual Stack Universal Platform Upgrade Kit
12 x 14 in.
304.8 mm
355.6 mm
Solaris 2000
1 Year
Certificate of Analysis (COA), Certificate of Conformance (COC)
12 in.
14 in.
Solaris 12x14 Dual Stack Universal platform – upgrade kit
One platform, screws, supporting rods