missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Solid Epoxy Work Surface

Labconco™ Solid Epoxy Work Surface for use with Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and Protector™ Work Stations.

Brand:  Labconco™ 3908401

Product Code. 15693587

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Molded solid epoxy work surface
  • Chemical-resistant and fire proof
  • Color: Black
  • Dished to contain spills
  • Contoured to fit dimensions of Protector Demonstration Hoods, Protector XVS Ventilation Systems Protector Work Stations
  • No cupsink cutout
  • Offered in 2 ft., 3 ft., 4 ft., 5 ft., 6 ft. and 8 ft. widths and 67.8cm (26.7 in.) and 90.1cm (35.5 in.) depths
Specifications

Specifications

Work Surface
26.7 in.
Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and Protector™ Work Stations
2.5 m
72 lb.
36 in.
Black
68 cm
1.0 in.
One year
33 kg
91 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.