missing translation for 'onlineSavingsMsg'
Learn More
Solketal, 97%, Thermo Scientific Chemicals Product Code.: 11347855

Solketal, 97%, Thermo Scientific Chemicals

Product Code. 11347855
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11347855

Brand: Thermo Scientific Alfa Aesar L02814.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-79-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00063238
InChI Key RNVYQYLELCKWAN-UHFFFAOYSA-N
Synonym solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketalShow MoreShow Less
PubChem CID 7528
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Melting Point -27°C
Density 1.064
Boiling Point 188°C to 190°C
Flash Point 80°C (176°F)
Refractive Index 1.434
Quantity 500 g
Beilstein 104465
Merck Index 14,5213
Solubility Information Soluble in water (fully),alcohol,acetals,esters,and ethers.
Formula Weight 132.16
Percent Purity 97%
Chemical Name or Material Solketal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243788 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberL02814.22,L02814.0E,L02814.36
Lot Number10151125 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02814.22,L02814.0E,L02814.36
Lot Number10121127 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02814.22,L02814.0E,L02814.36
Lot Number10236338 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02814.22,L02814.0E,L02814.36
Lot Number10199494 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02814.22,L02814.0E,L02814.36
compliance-icons
Product Identifier
  • Solketal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Solketal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Solketal, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.