missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Solutions for perfectION™ Combined ISE

Recommended reference solution for replacing electrolyte in ion selective electrodes.

Brand:  METTLER TOLEDO™ 51344755

137.75 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 12681296

  • 145.00 € / 300mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Keep electrolyte levels high by refilling regularly – when the probe is submerged, the electrolyte level should always be higher than the sample level.
  • Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
  • Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

perfectION Electrolyte FIon Electrolyte F
perfectION ion electrolyte E, 5 x 60 mL
Combined ISE Solution for perfectION
perfectION Electrode: Nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • perfectION Ion Electrolyte F

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P202-Do not handle until all safety precautions have been read and understood.

Supplemental information
  • MIXTURE LIST-Contains : Ammonium Sulfate, Silver Chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.