Learn More
Ansell™ Solvex™ 37-655 Series Green Nitrile Immersion Gloves Product Code.: 12610862
Add to basket
Ansell™ Solvex™ 37-655 Series Green Nitrile Immersion Gloves
Product Code. 12610862 Shop All Ansell Products
8
Click to view available options
Size:
7
8
9
10
11
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12610862
Brand: Ansell™ 37655080
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Fully reusable, with an unequaled abrasion resistance, it provides superb comfort for the wearer.
  • Provides superb comfort for the wearer
  • Food Approved
  • No inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities
  • The sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Cut Resistance Rating Level - 0
Exterior Finish Smooth
Product Type Immersion Gloves
Size 8
Style 37-655
Thickness (Metric) 0.38 mm
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Unflocked
Quantity 12 Pairs
Resistant To Chemical resistance
Series 37-655
Product Title
Ansell™ Solvex™ 37-655 Series Green Nitrile Immersion Gloves >