missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sonicated DNAs for Hybridization Studies

Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.

508.00 € - 613.00 €

Specifications

Format Solid
Content And Storage -20°C
Quantity 100/Pk.
For Use With (Application) Hybridisation experiments
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10723217
View Documents
Cytiva
27-4563-01
Sonicated Calf Thymus DNA
613.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10733217
View Documents
Cytiva
27-4565-01
Sonicated Salmon Sperm DNA
508.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp
Specifications

Specifications

Solid
-20°C
100/Pk.
Hybridisation experiments
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.