Learn More
Qsonica Q800R3 Sonicator
DNA Shearing Sonicator
Brand: Qsonica Q800R3
This item is not returnable.
View return policy
Description
Q800R3 is a complete package including a chiller to control temperature, digital operating system with programmable memory, high intensity ultrasonics and sound reducing enclosure.- Samples can be sheared to a range of fragment sizes (150bp – 3kb)
- System is compatible with commercially available sample tubes and is capable of processing up to 18 samples at one time
- Multiple tube rack options can accommodate 50μL to 1ml sample volumes using a variety of standard sample tubes
Specifications
DNA and Chromatin Shearing System | |
20 kHz | |
15.25 x 8 x 8.5 in. | |
Generator, Converter, Ultrasonic Horn, Rotation System, Chiller, Cables, Tubing, 12 x 0.5mL tube rack, 2-year warranty |
110 V | |
750 W | |
DNA shearing for Next-gen sequencing, Chromatin shearing, ChIP, ChIP-seq, RNA-seq, Protein extraction, Cell lysis | |
1 sec. to 10 hrs. |
Product Content Correction
Product Title
Qsonica Q800R3 Sonicator > Q800R3; 110V, 50/60Hz
Spot an opportunity for improvement?Share a Content Correction