Learn More
Syngene™ Sony™ A6 Gloss Paper
High density gloss print media with proper width for use in the Sony™ A6 Digital Black and White Thermal Printer.
Brand: Syngene™ UPP-110HG
This item is not returnable.
View return policy
Description
The UP-D898MD is a successor to the popular Sony™ UP-897 series of medical printers. Redesigned and upgraded, it offers several operational and ergonomic enhancements while retaining the same space-saving footprint and easy integration into medical carts.
- Enhanced usability with enlarged LCD display panel and joystick for intuitive menu navigation
- Delivers high quality black and white prints in under two seconds
- Advanced thermal printing technology delivers near-photo quality results using widely available print media
- Unique Sony™ technology allows paper to be cut in two directions – both up and down – as a lower cutter is newly included, along with the conventional upper cutter
- Flexible printer installation in almost any location, as well as smooth printing operations
- High quality thermal print engine can produce hard copy prints of still images captured for use in patient records and referrals
- Type V glossy paper
Specifications
Gloss Paper | |
A6 Digital Black and White Thermal Printer | |
20 m x 110 mm |